Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130213.1 | NP_001123685.1 | 4 - 5 | 793 | 69 | |
XM_005655370.3 | XP_005655427.1 | 569 | 69 | |||
XM_013988076.2 | XP_013843530.1 | 1451 | 69 | |||
XM_021088737.1 | XP_020944396.1 | 846 | 69 | |||
GenBank mRNA | AB530150.1 | - | 2 - 3 | 254 | 69 | |
AK237831.1 | - | 3 - 4 | 549 | 69 | ||
AK394594.1 | - | 3 - 4 | 554 | 69 | ||
AY429474.1 | - | 3 - 4 | 528 | 69 | ||
DQ866834.1 | - | 4 - 5 | 793 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |