Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005655370.3 | XP_005655427.1 | 1190 | 55 | ||
XM_013988076.2 | XP_013843530.1 | 2072 | 55 | |||
XM_021088737.1 | XP_020944396.1 | 1467 | 55 | |||
GenBank mRNA | AB530150.1 | - | 7 - 8 | 875 | 55 | |
AK237831.1 | - | 8 - 9 | 1170 | 55 | ||
AK394594.1 | - | 8 - 9 | 1175 | 55 | ||
AY429474.1 | - | 8 - 9 | 1149 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |