Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY669080.1 | - | 1 | 47 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |