Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204283.1 | NP_001191212.1 | 5 - 6 | 578 | 66 | |
GenBank mRNA | AK232196.1 | - | 5 - 6 | 578 | 66 | |
AK233807.1 | - | 5 - 6 | 552 | 66 | ||
AK394418.1 | - | 5 - 6 | 585 | 66 | ||
D50110.1 | - | 4 - 5 | 472 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |