Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005671280.3 | XP_005671337.1 | 1689 | 88 | ||
GenBank mRNA | AK346432.1 | - | 2 | 911 | 88 | |
DQ120777.1 | - | 1 | 743 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |