Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128454.1 | NP_001121926.1 | 3 - 4 | 371 | 77 | |
XM_013997036.2 | XP_013852490.1 | 707 | 77 | |||
GenBank mRNA | EU582021.1 | - | 3 - 4 | 371 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |