Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128467.1 | NP_001121939.1 | 3 - 4 | 638 | 106 | |
XM_021090894.1 | XP_020946553.1 | 817 | 106 | |||
GenBank mRNA | EU620575.1 | - | 3 - 4 | 638 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |