Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168416.1 | NP_001161888.1 | 3 | 277 | 74 | |
XM_013996999.2 | XP_013852453.1 | 445 | 74 | |||
GenBank mRNA | AB530141.1 | - | 3 | 199 | 74 | |
AK232124.1 | - | 3 | 252 | 74 | ||
AK395822.1 | - | 3 | 274 | 74 | ||
AY609880.1 | - | 3 | 255 | 74 | ||
L77214.1 | - | 3 | 154 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |