Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | M26235.1 | - | 3 | 656 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |