Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CILP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348903.1 | NP_001335832.1 | 9 | 68 | ||
XM_021083365.1 | XP_020939024.1 | 68 | ||||
GenBank mRNA | AK389521.1 | - | 1 | 68 | ||
U83114.1 | - | 1 | 1751 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |