Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ440724.1 | - | 2 | 241 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |