Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY312065.1 | - | 1 | 49 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Amplicon spans exons and probe does not span exons |