Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021062337.1 | XP_020917996.1 | 605 | 78 | ||
GenBank mRNA | AY550064.1 | - | 2 | 50 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons |