Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086056.1 | XP_020941715.1 | 1155 | 80 | ||
GenBank mRNA | DQ991099.1 | - | 1 | 54 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |