Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001266.2 | NP_001001266.1 | 3 | 256 | 108 | |
GenBank mRNA | M29515.1 | - | 3 | 256 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Inventoried Both primers and probe map within a single exon |