Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001627.1 | NP_001001627.1 | 3 | 650 | 78 | |
GenBank mRNA | AK232405.1 | - | 3 | 658 | 78 | |
AY533206.1 | - | 3 | 650 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |