Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044599.1 | NP_001038064.1 | 3 | 656 | 150 | |
GenBank mRNA | AK235391.1 | - | 3 | 602 | 150 | |
DQ529285.1 | - | 3 | 656 | 150 | ||
DQ665368.1 | - | 2 | 296 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Endogenous Controls Sus scrofa Inventoried Amplicon spans exons and probe does not span exons |