Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214111.1 | NP_999276.1 | 5 | 1147 | 63 | |
GenBank mRNA | AF216651.1 | - | 5 | 1147 | 63 | |
AK394765.1 | - | 4 | 754 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |