Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214187.1 | NP_999352.1 | 4 | 3357 | 115 | |
XM_005673404.3 | XP_005673461.1 | 3249 | 115 | |||
XM_005673405.3 | XP_005673462.1 | 3185 | 115 | |||
XM_013985894.2 | XP_013841348.1 | 3252 | 115 | |||
XM_013985895.2 | XP_013841349.1 | 3374 | 115 | |||
XM_013985896.2 | XP_013841350.1 | 3307 | 115 | |||
XM_021079505.1 | XP_020935164.1 | 3555 | 115 | |||
XM_021079506.1 | XP_020935165.1 | 3552 | 115 | |||
XM_021079507.1 | XP_020935166.1 | 3268 | 115 | |||
XM_021079508.1 | XP_020935167.1 | 3286 | 115 | |||
XM_021079509.1 | XP_020935168.1 | 3271 | 115 | |||
GenBank mRNA | AB092975.1 | - | 4 | 3357 | 115 | |
AB258452.1 | - | 2 | 3128 | 115 | ||
EF583903.1 | - | 1 | 2976 | 115 | ||
GU013761.1 | - | 1 | 2976 | 115 | ||
GU936184.1 | - | 1 | 2976 | 115 | ||
GU936185.1 | - | 1 | 2976 | 115 | ||
KC860784.1 | - | 1 | 115 | |||
KF019635.1 | - | 1 | 2976 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |