Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214247.1 | NP_999412.1 | 7 | 1950 | 70 | |
XM_013993694.2 | XP_013849148.1 | 2579 | 70 | |||
XM_021083198.1 | XP_020938857.1 | 2652 | 70 | |||
GenBank mRNA | AJ011767.1 | - | 7 | 1950 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |