Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214375.1 | NP_999540.1 | 6 | 1153 | 77 | |
XM_005657914.2 | XP_005657971.1 | 1191 | 77 | |||
XM_005657915.2 | XP_005657972.1 | 1419 | 77 | |||
XM_005657916.3 | XP_005657973.1 | 1885 | 77 | |||
XM_013991001.2 | XP_013846455.1 | 1195 | 77 | |||
XM_013991003.2 | XP_013846457.1 | 1415 | 77 | |||
XM_013991004.2 | XP_013846458.1 | 744 | 77 | |||
XM_021079519.1 | XP_020935178.1 | 1256 | 77 | |||
XM_021079520.1 | XP_020935179.1 | 1221 | 77 | |||
XM_021079521.1 | XP_020935180.1 | 1028 | 77 | |||
XM_021079522.1 | XP_020935181.1 | 1103 | 77 | |||
XM_021079523.1 | XP_020935182.1 | 1325 | 77 | |||
XM_021079524.1 | XP_020935183.1 | 1219 | 77 | |||
XM_021079525.1 | XP_020935184.1 | 899 | 77 | |||
XM_021079526.1 | XP_020935185.1 | 1021 | 77 | |||
XM_021079528.1 | XP_020935187.1 | 799 | 77 | |||
XM_021079529.1 | XP_020935188.1 | 1333 | 77 | |||
GenBank mRNA | AJ275968.1 | - | 6 | 1177 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |