Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102473.1 | NP_001095943.1 | 4 | 351 | 85 | |
GenBank mRNA | AB331640.1 | - | 4 | 351 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |