Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123122.1 | NP_001116594.1 | 4 | 450 | 111 | |
GenBank mRNA | AK348019.1 | - | 4 | 370 | 111 | |
AK391948.1 | - | 4 | 371 | 111 | ||
AK394505.1 | - | 4 | 452 | 111 | ||
AY609643.1 | - | 4 | 410 | 111 | ||
EU408324.1 | - | 4 | 450 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |