Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123145.1 | NP_001116617.1 | 2 | 248 | 82 | |
XM_021097906.1 | XP_020953565.1 | 2669 | 82 | |||
XM_021097907.1 | XP_020953566.1 | 2250 | 82 | |||
GenBank mRNA | EU375564.1 | - | 2 | 248 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |