Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHEROC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123161.1 | NP_001116633.1 | 2 | 148 | 74 | |
GenBank mRNA | AK399215.1 | - | 2 | 206 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |