Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03388131_u1
See other IGF2 GE Assays ›
Gene Symbol:
IGF2
Gene Name:
insulin like growth factor 2
Gene Aliases:
IGF-II
Chromosome Location:
Chr.2: 1469183 - 1496417 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03388131_u1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: IGF2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_213883.2
NP_999048.1
4
649
85
XM_021080575.1
XP_020936234.1
1429
85
XM_021080576.1
XP_020936235.1
1423
85
XM_021080582.1
XP_020936241.1
543
85
XM_021080585.1
XP_020936244.1
719
85
XM_021080593.1
XP_020936252.1
318
85
XM_021080596.1
XP_020936255.1
1160
85
XM_021080603.1
XP_020936262.1
1111
85
XM_021080607.1
XP_020936266.1
871
85
XM_021080612.1
XP_020936271.1
377
85
XM_021080616.1
XP_020936275.1
433
85
XM_021080621.1
XP_020936280.1
1102
85
XM_021080632.1
XP_020936291.1
1097
85
XM_021080637.1
XP_020936296.1
424
85
XM_021080641.1
XP_020936300.1
361
85
XM_021080643.1
XP_020936302.1
649
85
XM_021080648.1
XP_020936307.1
648
85
GenBank mRNA
AF466299.1
-
3
375
85
AK232525.1
-
3
721
85
AK233225.1
-
4
636
85
AK233453.1
-
5
856
85
AK234422.1
-
3
728
85
AK239902.1
-
3
446
85
AK240457.1
-
3
398
85
AK394055.1
-
4
628
85
AK395456.1
-
3
723
85
AK395812.1
-
3
728
85
AK398925.1
-
5
828
85
AK400540.1
-
3
710
85
AK400765.1
-
3
566
85
DQ295330.1
-
2
166
85
HQ450752.1
-
4
597
85
HQ450753.1
-
5
790
85
HQ450754.1
-
5
829
85
HQ450755.1
-
3
684
85
HQ450756.1
-
3
356
85
HQ450757.1
-
3
380
85
HQ450758.1
-
3
323
85
JN120799.1
-
3
456
85
JN120800.1
-
4
612
85
X56094.1
-
649
85
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Recommended Assay
Sus scrofa
Amplicon spans exons and probe does not span exons
Panther Classification:
Molecular Function -
growth factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0