Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005152.2 | NP_001005152.1 | 1 - 2 | 645 | 92 | |
GenBank mRNA | AK239706.1 | - | 1 - 2 | 540 | 92 | |
AK240333.1 | - | 1 - 2 | 669 | 92 | ||
AK350394.1 | - | 1 - 2 | 669 | 92 | ||
AK398037.1 | - | 1 - 2 | 703 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |