Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044614.2 | NP_001038079.1 | 2 - 3 | 584 | 111 | |
GenBank mRNA | AK389176.1 | - | 2 - 3 | 598 | 111 | |
AK395712.1 | - | 2 - 3 | 625 | 111 | ||
AK395910.1 | - | 3 - 4 | 679 | 111 | ||
AK396682.1 | - | 1 - 2 | 1109 | 111 | ||
AK400522.1 | - | 2 - 3 | 596 | 111 | ||
DQ365929.1 | - | 2 - 3 | 585 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |