Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077226.1 | NP_001070694.1 | 2 - 3 | 254 | 147 | |
XM_005664070.3 | XP_005664127.1 | 459 | 147 | |||
XM_013997827.2 | XP_013853281.1 | 1418 | 147 | |||
GenBank mRNA | AK348596.1 | - | 4 - 5 | 416 | 147 | |
AK396389.1 | - | 4 - 5 | 413 | 147 | ||
DQ972959.1 | - | 2 - 3 | 254 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |