Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077228.1 | NP_001070696.1 | 5 - 6 | 519 | 83 | |
XM_021077335.1 | XP_020932994.1 | 641 | 83 | |||
XM_021077336.1 | XP_020932995.1 | 774 | 83 | |||
XM_021077337.1 | XP_020932996.1 | 655 | 83 | |||
XM_021077338.1 | XP_020932997.1 | 566 | 83 | |||
XM_021077339.1 | XP_020932998.1 | 649 | 83 | |||
XM_021077340.1 | XP_020932999.1 | 567 | 83 | |||
GenBank mRNA | AK231168.1 | - | 7 - 8 | 781 | 83 | |
AY609388.1 | - | 6 - 7 | 748 | 83 | ||
DQ972968.1 | - | 5 - 6 | 519 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |