Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097481.2 | NP_001090950.1 | 2 - 3 | 202 | 91 | |
GenBank mRNA | AK237453.1 | - | 2 - 3 | 202 | 91 | |
AK392786.1 | - | 2 - 3 | 203 | 91 | ||
AK399720.1 | - | 2 - 3 | 204 | 91 | ||
AY610367.1 | - | 1 - 2 | 183 | 91 | ||
AY610469.1 | - | 1 - 2 | 185 | 91 | ||
DQ629173.1 | - | 2 - 3 | 203 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |