Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097493.1 | NP_001090962.1 | 6 - 7 | 830 | 78 | |
XM_005660185.3 | XP_005660242.1 | 2478 | 78 | |||
XM_021096459.1 | XP_020952118.1 | 979 | 78 | |||
XM_021097074.1 | XP_020952733.1 | 980 | 78 | |||
GenBank mRNA | AK231194.1 | - | 3 - 4 | 914 | 78 | |
AK348760.1 | - | 6 - 7 | 951 | 78 | ||
DQ439546.1 | - | 6 - 7 | 830 | 78 | ||
DQ439547.1 | - | 6 - 7 | 830 | 78 | ||
DQ439548.1 | - | 1 - 2 | 40 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |