Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097516.1 | NP_001090985.1 | 3 - 4 | 318 | 61 | |
GenBank mRNA | AK238737.1 | - | 3 - 4 | 342 | 61 | |
AK346167.1 | - | 3 - 4 | 321 | 61 | ||
AK389782.1 | - | 3 - 4 | 332 | 61 | ||
AK400044.1 | - | 3 - 4 | 378 | 61 | ||
AY682219.1 | - | 3 - 4 | 349 | 61 | ||
AY682222.1 | - | 3 - 4 | 349 | 61 | ||
AY682832.1 | - | 3 - 4 | 335 | 61 | ||
DQ486897.1 | - | 3 - 4 | 318 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |