Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYD88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099923.1 | NP_001093393.1 | 4 - 5 | 737 | 133 | |
GenBank mRNA | AB292176.1 | - | 4 - 5 | 774 | 133 | |
EF198416.1 | - | 4 - 5 | 737 | 133 | ||
EU056736.1 | - | 4 - 5 | 133 | |||
EU077229.1 | - | 4 - 5 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |