Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101033.1 | NP_001094503.1 | 15 - 16 | 1506 | 86 | |
XM_005672829.3 | XP_005672886.1 | 3005 | 86 | |||
GenBank mRNA | AK237864.1 | - | 16 - 17 | 1512 | 86 | |
AK350647.1 | - | 15 - 16 | 1523 | 86 | ||
DQ859019.1 | - | 15 - 16 | 1506 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |