Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110173.1 | NP_001103643.1 | 2 - 3 | 316 | 65 | |
GenBank mRNA | AK349078.1 | - | 2 - 3 | 630 | 65 | |
EU165323.1 | - | 2 - 3 | 316 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |