Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110429.1 | NP_001103899.1 | 2 - 3 | 400 | 105 | |
XM_005665984.3 | XP_005666041.1 | 423 | 105 | |||
XM_005665985.3 | XP_005666042.1 | 378 | 105 | |||
XM_013977923.2 | XP_013833377.1 | 407 | 105 | |||
GenBank mRNA | AK234045.1 | - | 2 - 3 | 377 | 105 | |
AK346620.1 | - | 2 - 3 | 340 | 105 | ||
KU705641.1 | - | 1 - 2 | 271 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |