Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113039.2 | NP_001106510.2 | 1 - 2 | 339 | 95 | |
NM_001293317.1 | NP_001280246.1 | 1 - 2 | 339 | 95 | ||
NR_120680.1 | 1 - 2 | 339 | 95 | |||
GenBank mRNA | AB188301.2 | - | 1 - 2 | 210 | 95 | |
AJ628065.1 | - | 1 - 2 | 97 | 95 | ||
AK230475.1 | - | 1 - 2 | 185 | 95 | ||
AK230585.1 | - | 1 - 2 | 188 | 95 | ||
AK389696.1 | - | 1 - 2 | 308 | 95 | ||
AK396573.1 | - | 1 - 2 | 339 | 95 | ||
AY535422.1 | - | 1 - 2 | 91 | 95 | ||
DB797683.1 | - | 1 - 2 | 188 | 95 | ||
EU716413.1 | - | 1 - 2 | 91 | 95 | ||
FJ555286.1 | - | 1 - 2 | 91 | 95 | ||
FJ555287.1 | - | 1 - 2 | 91 | 95 | ||
GQ304754.1 | - | 1 - 2 | 91 | 95 | ||
GU723684.1 | - | 1 - 2 | 91 | 95 | ||
GU723686.1 | - | 1 - 2 | 91 | 95 | ||
KF460453.1 | - | 1 - 2 | 132 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |