Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113217.1 | NP_001106688.1 | 4 - 5 | 475 | 88 | |
GenBank mRNA | AK343747.1 | - | 4 - 5 | 460 | 88 | |
AK343823.1 | - | 4 - 5 | 451 | 88 | ||
AK351256.1 | - | 4 - 5 | 452 | 88 | ||
AY550040.1 | - | 1 - 2 | 79 | 88 | ||
AY735454.1 | - | 4 - 5 | 475 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |