Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113697.1 | NP_001107169.1 | 3 - 4 | 477 | 134 | |
XM_013985516.2 | XP_013840970.1 | 409 | 134 | |||
GenBank mRNA | AK348355.1 | - | 3 - 4 | 342 | 134 | |
AK349578.1 | - | 3 - 4 | 399 | 134 | ||
AY609503.1 | - | 4 - 5 | 434 | 134 |
Gene Symbol: LOC100525472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666531.3 | XP_005666588.1 | 317 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |