Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113699.1 | NP_001107171.1 | 1 - 2 | 1030 | 83 | |
XM_005673033.3 | XP_005673090.1 | 1841 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |