Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113704.1 | NP_001107176.1 | 4 - 5 | 1017 | 156 | |
GenBank mRNA | AF464007.1 | - | 4 - 5 | 1017 | 156 | |
AF464008.1 | - | 4 - 5 | 1017 | 156 | ||
AF464019.1 | - | 4 - 5 | 1017 | 156 | ||
AF464020.2 | - | 4 - 5 | 979 | 156 | ||
AK236916.1 | - | 3 - 4 | 737 | 156 | ||
AK239694.1 | - | 4 - 5 | 998 | 156 | ||
AK348801.1 | - | 3 - 4 | 726 | 156 | ||
AY247772.1 | - | 4 - 5 | 1017 | 156 | ||
AY459304.1 | - | 4 - 5 | 1017 | 156 | ||
AY463538.1 | - | 1 - 2 | 136 | 156 | ||
DQ883214.1 | - | 4 - 5 | 1015 | 156 | ||
DQ883216.1 | - | 4 - 5 | 1017 | 156 | ||
DQ976363.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992502.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992503.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992504.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992505.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992506.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992507.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992508.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992509.1 | - | 4 - 5 | 1005 | 156 | ||
DQ992510.1 | - | 4 - 5 | 1005 | 156 | ||
EU432054.1 | - | 4 - 5 | 998 | 156 | ||
EU432055.1 | - | 4 - 5 | 998 | 156 | ||
EU432056.1 | - | 4 - 5 | 998 | 156 | ||
EU432057.1 | - | 4 - 5 | 998 | 156 | ||
GU322913.1 | - | 3 - 4 | 707 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |