Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114286.1 | NP_001107758.1 | 1 - 2 | 122 | 100 | |
GenBank mRNA | EF682141.1 | - | 1 - 2 | 122 | 100 | |
KF500033.1 | - | 1 - 2 | 210 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |