Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122995.2 | NP_001116467.1 | 2 - 3 | 354 | 114 | |
NM_001301326.1 | NP_001288255.1 | 2 - 3 | 351 | 114 | ||
GenBank mRNA | AK392493.1 | - | 2 - 3 | 351 | 114 | |
BP172930.1 | - | 2 - 3 | 354 | 114 | ||
CK454400.1 | - | 1 - 2 | 170 | 114 | ||
EU545196.1 | - | 2 - 3 | 184 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |