Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123043.1 | NP_001116515.1 | 2 - 3 | 372 | 54 | |
XM_005665245.3 | XP_005665302.1 | 561 | 54 | |||
GenBank mRNA | AK390132.1 | - | 3 - 4 | 508 | 54 | |
AK400244.1 | - | 3 - 4 | 540 | 54 | ||
DQ917651.1 | - | 2 - 3 | 372 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |