Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123089.2 | NP_001116561.1 | 6 - 7 | 759 | 71 | |
GenBank mRNA | EU076594.1 | - | 6 - 7 | 760 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |