Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123119.1 | NP_001116591.1 | 3 - 4 | 738 | 55 | |
XM_005665820.3 | XP_005665877.2 | 751 | 55 | |||
GenBank mRNA | EU282365.1 | - | 3 - 4 | 463 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |