Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123131.1 | NP_001116603.1 | 5 - 6 | 721 | 76 | |
GenBank mRNA | AK237361.1 | - | 5 - 6 | 687 | 76 | |
AK392188.1 | - | 5 - 6 | 933 | 76 | ||
AK398775.1 | - | 5 - 6 | 824 | 76 | ||
DQ917621.1 | - | 5 - 6 | 721 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |