Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123158.1 | NP_001116630.1 | 7 - 8 | 1374 | 71 | |
XM_005673043.3 | XP_005673100.1 | 1418 | 71 | |||
GenBank mRNA | AK232604.1 | - | 2 - 3 | 310 | 71 | |
AK345870.1 | - | 7 - 8 | 1372 | 71 | ||
AK346076.1 | - | 7 - 8 | 1342 | 71 | ||
AK393110.1 | - | 7 - 8 | 1338 | 71 | ||
AK393671.1 | - | 7 - 8 | 1338 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |