Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123208.1 | NP_001116680.1 | 6 - 7 | 867 | 61 | |
XM_013977549.2 | XP_013833003.2 | 2118 | 61 | |||
XM_021097926.1 | XP_020953585.1 | 807 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |